Echtgrün FCF

Lieferant: NATIONAL DIAGNOSTICS
Achtung

Synonyms: Light green, Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)-5-hydroxybenzenesulphonate, CI 42053, Dihydrogen (ethyl)[4-[4-[ethyl(3-sulphonatobenzyl)amino](4-hydroxy-2-sulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene](3-sulphonatobenzyl)ammonium, disodium salt

HS-516-25G
NADIHS-516-25GEA 0 CHF
NADIHS-516-25G
Echtgrün FCF
Echtgrün FCF
Formel: C₃₇H₃₄N₂Na₂O₁₀S₃
MW: 808.86 g/mol
Melting Pt: 290
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013053
CAS-Nummer: 2353-45-9
EINECS: 219-091-5
Merck Index: 13,03970

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR