Mercaptoethanol

Lieferant: NATIONAL DIAGNOSTICS
Gefahr

Synonyms: 2-Hydroxyethyl mercaptan, β-Mercaptoethanol

EC-603-50ML
NADIEC-603-50MLEA 0 CHF
NADIEC-603-50ML
Mercaptoethanol
Mercaptoethanol
Formel: HSCH₂CH₂OH
MW: 78.14 g/mol
Siedepunkt: 157
Melting Pt: –100 °C
Dichte: 1.12 g/cm³ (20 °C)
Flash-Pt: 68
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004890
CAS-Nummer: 60-24-2
EINECS: 200-464-6
UN: 2966
ADR: 6.1 II
Merck Index: 13,05894
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc427209b84d0343a5ae47c435ddfc53df4013d2&vid=7d70fd087d948d54e7a946d3b2310e2d820d2396&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR