Inverse Mikroskope, AE2000
Lieferant: MOTIC
1101001402061 1100103800043 1100103800054 1101001703251 1101001703261 1101001703271 1101001703291 1101001703091 1101001703281 1101001703102 1101001703112 P2HD P1LED P2LED P1S P2S 1100103800088 1100103800139
630-1456EA 207 CHF
630-1456 630-1316 630-1317 630-1367 630-1368 630-1369 630-1371 630-1364 630-1370 630-1366 630-1501 630-2970 MOTIP1LED MOTIP2LED MOTIP1S MOTIP2S 630-3459 630-3460
Inverse Mikroskope, AE2000
Mikroskope
Die inversen Mikroskope AE2000 sind für Zellkulturanwendungen vorgesehen.
  • Automatic turn off after 15 m of non use
  • RoHs compliant lead-free objectives
  • Robust manufacturing with small footprint
Suitable for microbiology and clinical labs, especially for routine cell inspections.
Bestellinformation: Special sets including microscope, application-specific accessories and three objectives (4×, Ph 10×, LWD Ph 20×) are available as well.
Lieferumfang: Additionnal phase contrast objectives are also available. LED modules can be ordered separately.