4-Chlorphenylcyclopropylketon

Lieferant: Molekula

Synonyms: (4-Chlorophenyl)(cyclopropyl)methanone

M86903619
MOLEM86903619EA 0 CHF
MOLEM86903619
4-Chlorphenylcyclopropylketon
4-Chlorphenylcyclopropylketon
Formel: C₁₀H₉ClO
MW: 180,63 g/mol
MDL Number: MFCD00001295
CAS-Nummer: 6640-25-1
EINECS: 229-655-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR