L(+)-Asparagin Monohydrat

Lieferant: Molekula

Synonyms: L-Asparagine monohydrate, H-Asn-OH·H₂O, (S)-(+)-Asparagine monohydrate, (S)-2,4-Diamino-4-oxobutanoic acid hydrate, H-Asn-OH.H2O

M86033702
MOLEM86033702EA 0 CHF
MOLEM86033702
L(+)-Asparagin Monohydrat
L(+)-Asparagin Monohydrat
Formel: C₄H₈N₂O₃·1H₂O
MW: 150.13 g/mol
Melting Pt: 215…217 °C
Dichte: 1.567 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00151038
CAS-Nummer: 5794-13-8
EINECS: 200-735-9
Merck Index: 14,00837

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR