Diisopropylcarbodiimid

Lieferant: Molekula

Synonyms: N,N'-methanetetraylbis(1-methylethylamine), N,N-Diisopropylcarbodiimide, 1,3-Diisopropylcarbodiimide, N,N'-Diisopropylcarbodiimide, N.N’-Diisopropylcarbodiimide, DIC

M79451433
MOLEM79451433EA 0 CHF
MOLEM79451433
Diisopropylcarbodiimid
Diisopropylcarbodiimid
Formel: C₇H₁₄N₂
MW: 126,2 g/mol
Siedepunkt: 145 °C (1013 hPa)
Dichte: 0.81 g/cm³ (20 °C)
Flash-Pt: 33 °C
MDL Number: MFCD00065689
CAS-Nummer: 693-13-0
EINECS: 211-743-7
UN: 2929
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR