Purpurin (1,2,4-Trihydroxy-9,10-anthrachinon)

Lieferant: Molekula

M65000587
MOLEM65000587EA 0 CHF
MOLEM65000587
Purpurin (1,2,4-Trihydroxy-9,10-anthrachinon)
Purpurin (1,2,4-Trihydroxy-9,10-anthrachinon)
Formel: C₁₄H₈O₅
MDL Number: MFCD00001203
CAS-Nummer: 81-54-9
EINECS: 201-359-8

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR