4-(Hydroxymethyl)phenylboronsäure

Lieferant: Molekula

Synonyms: 4-(Hydroxymethyl)benzeneboronic acid

M54140413
MOLEM54140413EA 0 CHF
MOLEM54140413
4-(Hydroxymethyl)phenylboronsäure
4-(Hydroxymethyl)phenylboronsäure
Formel: C₇H₉BO₃
MW: 151,96 g/mol
MDL Number: MFCD00792672
CAS-Nummer: 59016-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR