Tannin

Lieferant: Molekula

Synonyms: Gallotannin, Tannin

M46862937
MOLEM46862937EA 0 CHF
MOLEM46862937
Tannin
Tannin
Formel: C₇₆H₅₂O₄₆
MW: 1701.2 g/mol
Melting Pt: 218…219 °C
Dichte: 1 g/cm³ (20 °C)
Flash-Pt: 199
MDL Number: MFCD00066397
CAS-Nummer: 1401-55-4
EINECS: 215-753-2
Merck Index: 12,09221

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR