3H-1,2-Benzodithiol-on-1,1-dioxid (Beaucage-Reagenz)

Lieferant: Molekula

Synonyms: Beaucage Reagent, 3H-1,2-Benzodithiol-3-one-1,1-dioxide, 3H-1,2-Benzodithiol-3-one 1,1-Dioxide

M46641343
MOLEM46641343EA 0 CHF
MOLEM46641343
3H-1,2-Benzodithiol-on-1,1-dioxid (Beaucage-Reagenz)
3H-1,2-Benzodithiol-on-1,1-dioxid (Beaucage-Reagenz)
Formel: C₇H₄O₃S₂
MW: 200,24 g/mol
MDL Number: MFCD00132960
CAS-Nummer: 66304-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR