Sarkosin

Lieferant: Molekula

Synonyms: H-Sar-OH, N-Methylglycine, Methylaminoacetic acid

M29103605
MOLEM29103605EA 0 CHF
MOLEM29103605
Sarkosin
Sarkosin
Formel: C₃H₇NO₂
MW: 89,09 g/mol
Siedepunkt: 185 °C (1013 hPa)
Melting Pt: 63 °C
Dichte: 1.301 g/cm³ (20 °C)
MDL Number: MFCD00004279
CAS-Nummer: 107-97-1
EINECS: 203-538-6
Merck Index: 12,08519

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR