1-(3,4-Dichlorphenyl)piperazin

Lieferant: Molekula
Achtung

M27310081
MOLEM27310081EA 0 CHF
MOLEM27310081
1-(3,4-Dichlorphenyl)piperazin
1-(3,4-Dichlorphenyl)piperazin
Formel: C₁₀H₁₂Cl₂N₂
MW: 231.12 g/mol
Siedepunkt: 370
Melting Pt: 62…65 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00023128
CAS-Nummer: 57260-67-0
EINECS: 260-652-9

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR