Ethidiumbromid

Lieferant: Molekula
Gefahr

Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide

M16559763
MOLEM16559763EA 0 CHF
MOLEM16559763
Ethidiumbromid
Ethidiumbromid
Formel: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
Siedepunkt: 72 °C (0.1 mmHg)
Melting Pt: 261…264 °C
Dichte: 1.01 g/cm³ (20 °C)
Flash-Pt: <100 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011724
CAS-Nummer: 1239-45-8
EINECS: 214-984-6
UN: 2811
ADR: 6.1 I
Merck Index: 13,04751

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR