Trioctylphosphin

Lieferant: Molekula

Synonyms: TOP, Tri-n-octylphosphine

73693902-25G
MOLE73693902-25GEA 0 CHF
MOLE73693902-25G
Trioctylphosphin
Trioctylphosphin
Formel: C₂₄H₅₁P
MW: 370.64 g/mol
Siedepunkt: 150…155 °C (0.03 mmHg)
Melting Pt: 48
Dichte: 0.831 g/cm³ (20 °C)
MDL Number: MFCD00015298
CAS-Nummer: 4731-53-7
EINECS: 225-234-2
UN: 1760
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR