3-Nitro-p-toluylsäure

Lieferant: Molekula

Synonyms: 4-Methyl-3-nitrobenzoic acid

41708041-100G
MOLE41708041-100GEA 0 CHF
MOLE41708041-100G
3-Nitro-p-toluylsäure
3-Nitro-p-toluylsäure
Formel: C₈H₇NO₄
MW: 181,15 g/mol
Siedepunkt: 360 °C (1013 hPa)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007174
CAS-Nummer: 96-98-0
EINECS: 202-549-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR