2-Amino-1,3-propandiol

Lieferant: Molekula

Synonyms: Serinol, 2-Amino-1,3-dihydroxy propane, 2-Aminopropane-1,3-diol

39331277-5G
MOLE39331277-5GEA 0 CHF
MOLE39331277-5G
2-Amino-1,3-propandiol
2-Amino-1,3-propanediol
Formel: C₃H₉NO₂
Siedepunkt: 267 °C (1013 hPa)
Melting Pt: 91…95 °C
MDL Number: MFCD00051487
CAS-Nummer: 534-03-2
EINECS: 208-584-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR