Tetramethylammoniumbromid

Lieferant: Molekula
Gefahr

38302697-100G
MOLE38302697-100GEA 0 CHF
MOLE38302697-100G
Tetramethylammoniumbromid
Tetramethylammonium bromide
Formel: C₄H₁₂BrN
MW: 154.05 g/mol
Siedepunkt: ∼360 °C (1013 hPa)
Dichte: 1.56 g/cm³ (20 °C)
MDL Number: MFCD00011626
CAS-Nummer: 64-20-0
EINECS: 200-581-2
UN: 2811
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR