1-Aminopyridiniumiodid

Lieferant: Molekula

Synonyms: 1-Aminopyridinium Iodide, 1-Aminopyridine hydroiodide, 1-Aminopyridine HI

37769177-5G
MOLE37769177-5GEA 0 CHF
MOLE37769177-5G
1-Aminopyridiniumiodid
1-Aminopyridinium iodide
Formel: C₅H₇IN₂
MW: 222.03 g/mol
Melting Pt: 161…162 °C
MDL Number: MFCD00051893
CAS-Nummer: 6295-87-0
EINECS: 228-566-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR