2-Chlor-5-(trifluormethyl)pyridin

Lieferant: Molekula

Synonyms: 2-Chloro-5-trifluoromethylpyridine

34032938-25G
MOLE34032938-25GEA 0 CHF
MOLE34032938-25G
2-Chlor-5-(trifluormethyl)pyridin
2-Chloro-5-(trifluoromethyl)pyridine
Formel: C₆H₃ClF₃N
Siedepunkt: 152 °C (1013 hPa)
Melting Pt: 30…31 °C
Dichte: 1.41 g/cm³ (20 °C)
MDL Number: MFCD00042225
CAS-Nummer: 52334-81-3
EINECS: 257-856-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR