Oxytetracyclin Hydrochlorid

Lieferant: Molekula

29241094-25G
MOLE29241094-25GEA 0 CHF
MOLE29241094-25G
Oxytetracyclin Hydrochlorid
Oxytetracyclin Hydrochlorid
Formel: C₂₂H₂₅ClN₂O₉
MW: 496.90 g/mol
MDL Number: MFCD00135815
CAS-Nummer: 2058-46-0
EINECS: 218-161-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR