Brij® 35, Pulver

Lieferant: Molekula

Synonyms: Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23

28156962-1KG
MOLE28156962-1KGEA 0 CHF
MOLE28156962-1KG
Brij® 35, Pulver
Brij® 35
Formel: C₅₈H₁₁₈O₂₄
MW: 1199.56 g/mol
MDL Number: MFCD00080891
CAS-Nummer: 9002-92-0
EINECS: 500-002-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR