2-Picolylchlorid Hydrochlorid

Lieferant: Molekula

Synonyms: 2-(Chloromethyl)pyridine HCl, 2-(Chloromethyl)pyridine hydrochloride

25430297-5G
MOLE25430297-5GEA 0 CHF
MOLE25430297-5G
2-Picolylchlorid Hydrochlorid
2-Picolylchlorid Hydrochlorid
Formel: C₆H₇Cl₂N
MW: 164.03 g/mol
MDL Number: MFCD00012811
CAS-Nummer: 6959-47-3
EINECS: 230-149-9
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR