Terephthalaldehyd

Lieferant: Molekula

Synonyms: Terephthaldialdehyde, Terephthaldicarboxaldehyde, p-Phthalaldehyde

23638396-100G
MOLE23638396-100GEA 0 CHF
MOLE23638396-100G
Terephthalaldehyd
Terephthalaldehyd
Formel: C₈H₆O₂
Siedepunkt: 245 °C (1013 hPa)
Dichte: 1.381 g/cm³ (20 °C)
MDL Number: MFCD00006949
CAS-Nummer: 623-27-8
EINECS: 210-784-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR