Gummi Arabicum

Lieferant: Molekula
Achtung

Synonyms: Acacia, Acacia gum

13599707-100G
MOLE13599707-100GEA 0 CHF
MOLE13599707-100G
Gummi Arabicum
Gummi Arabicum
Dichte: 1,35 g/cm³ (20 °C)
Flash-Pt: >250 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00081264
CAS-Nummer: 9000-01-5
EINECS: 232-519-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR