Tetrabutylammoniumhydroxid 40% aqueous solution

Lieferant: Molekula
Gefahr

12269822-100ML
MOLE12269822-100MLEA 0 CHF
MOLE12269822-100ML
Tetrabutylammoniumhydroxid 40% aqueous solution
Tetrabutylammoniumhydroxid
Formel: C₁₆H₃₇NO
MW: 259.48 g/mol
Melting Pt: 29.85
Dichte: 0.968
Lager Temperatur: Kühlschrank
MDL Number: MFCD00009425
CAS-Nummer: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR