Sponge-Sticks
Lieferant: 3M Food Safety
SSL10BPW SSL100 SSL10NB SSL10LET
MMMPSSL10BPWEA 324 CHF
MMMPSSL10BPW MMMPSSL100 MMMPSSL10NB MMMPSSL10LET
Sponge-Sticks
Tupfer Tupfer für die Probenahme
3M™ Sponge-Stick in sample bag with 10 ml D/E neutralising broth uses simplified packaging to minimise handling and preparation, and reduces waste.
  • Stick-mounted sponge design allows the ability to access hard to reach areas without directly handling the sponge
  • Stick-mounted sponge design provides the ability to easily snap off the sponge from the handle after swabbing
  • Simplified packaging minimises handling and preparation and reduces waste
Better testing begins with better samples and better samples are obtained when you use the best collectionMore Product Information