Hydrated sponge
Lieferant: 3M Food Safety
HS10BPW HS10BPW2G HS10LET
MMMPHS10BPWEA 290 CHF
MMMPHS10BPW MMMPHS10BPW2G MMMPHS10LET
Hydrated sponge
Tupfer Tupfer für die Probenahme
3M™ Hydrated-Sponge uses simplified packaging to minimise handling and preparation, and reduces waste.
  • Ready-to-use, pre-hydrated sponges for easy and convenient environmental and food sample collection
  • Durable sponge withstands scrubbing
  • Sponge bag with write-on label for ease in labelling samples
  • Includes two decontaminated gloves for safety and convenience
This biocide-free cellulose sponge is pre-hydrated with diluent for convenient sample collection. The biocide-free cellulose sponge does not interfere with microorganism viability within 48 hoursMore Product Information