Foam insert for 3M™ Solus™ 1000 spectacles
Lieferant: 3M
SOLUSSR
MMMASOLUSSREA 52.9 CHF
MMMASOLUSSR
Foam insert for 3M™ Solus™ 1000 spectacles
Schutzbrillen
Foam insert designed for use with 3M™ Solus™ 1000 safety spectacles.
  • Fills the gap between the eyewear and the wearer's face, further reducing the risk of injury from projectiles and other debris
  • Offers extra comfort and protection from sweat, ideal for hot and humid conditions
Order Now
TECHNISCHE DATEN
Environmentally Preferable