Nailsaver Scheuerschwämme
Lieferant: 3M
SCH105 SCH106 SCH363 SCH108 SCH107 SCH2276
MMMASCH105EA 41.9 CHF
MMMASCH105 MMMASCH106 MMMASCH363 MMMASCH108 MMMASCH107 MMMASCH2276
Nailsaver Scheuerschwämme
Wischtücher Schwämme
3M™ Nailsaver scouring sponges are cleaning sponges made of a textured synthetic fibre white web containing very soft abrasive particles, laminated with an adhesive onto a grooved synthetic colour-coded foam holder.
  • Easy positioning four colours of foams) to comply with HACCP requirements
  • Good cleaning performance for light-duty cleaning tasks
  • Soft and low scratching power on most surfaces
  • Easy to handle and finger protection thanks to the shape of the foam
  • Very long lasting
Verwenden Sie für eine einfache Reinigung mit geringer Kratzkraft auf denMore Product Information