Microplate reader, SpectraMax® L
Lieferant: Molecular Devices
SPECTRALCONFIG 0310-5628 0310-5126 0310-5649 0310-5882 0310-5650 0310-5694
MLDVSPECTRALCONFIGEA 0 CHF
MLDVSPECTRALCONFIG MLDV0310-5628 MLDV0310-5126 MLDV0310-5649 MLDV0310-5882 MLDV0310-5650 MLDV0310-5694
Microplate reader, SpectraMax® L
Mikrotiterplatten-Photometer
The SpectraMax® L microplate reader measures flash and glow luminescence assays in both 96- and 384-well plates.
  • Detect dim samples with unsurpassed sensitivity
  • Avoid saturation issues with 9 orders of dynamic range
  • Enable robust performance with autorinse injectors
  • Ensure compliance with IQ/OQ and FDA CFR 21 Part 11 compliance tools
  • Easy set-up and analysis with industry-leading SoftMax Pro software
SpectraMax® L is upgradeable to include a barcode reader, programmable injectors, and dual-channel configurations for a 2-fold increase in mediumMore Product Information