Milli-Q® SQ 240CV Purification System
Lieferant: Merck
ZSQ240UCVT0
MILLZSQ240UCVT0EA 14040 CHF
MILLZSQ240UCVT0
Milli-Q® SQ 240CV Purification System
Wasseraufbereitungssysteme Reinstwassersysteme
The Milli-Q® SQ 240CV water purification system delivers both pure reverse osmosis (RO) water and ultrapure (Type 1) water. It benefits from a fixed preset volume dispenser that records and repeats the dispensing volume of ultrapure water.
  • High-flow (up to 42 L/h) RO cartridge rapidly refills (as few as 6 min) mobile SQ Switch tank with freshly produced pure RO water
  • Gravity Valve (optional) lets you dispense pure RO water at up to 6 L/h directly from the SQ Switch tank base
  • RO water recovery loop saves 50% of the rejected water
  • Automatic RO membrane