Millicell® EZ Slides
Lieferant: Merck
PEZGS0896 PEZGS0496 PEZGS0416 PEZGS0816
MILFPEZGS0896EA 923 CHF
MILFPEZGS0896 MILFPEZGS0496 MILFPEZGS0416 MILFPEZGS0816
Millicell® EZ Slides
Objektträger Mikroskop-Objektträger
Millicell® EZ Slide allows the user to grow, fix, and stain cells directly on a microscope slide. There is no need to disassemble the unit or transfer cells from one device to another prior to analysis.
The polypropylene wells are resistant to most fixatives used, so the cells can be grown on the slide and stained right in the well. Once stained, the microscope slide can be easily removed by pressing down the four side tabs of the unit and lifting the wells off of the slide.
This easy slide removal reduces the risk of damaging the cells and no special tools are required to remove theMore Product Information
Order Now
TECHNISCHE DATEN
Environmentally Preferable