Saures Reinigungs- und Neutralisationsmittel, ProCare Lab 30 P
Lieferant: MIELE
10266500 10266490
MIEL10266500EA 131 CHF
MIEL10266500 MIEL10266490
Saures Reinigungs- und Neutralisationsmittel, ProCare Lab 30 P
Reinigungsmittel Waschmittel und Oberflächenreiniger
Liquid, acidic cleaning concentrate for automated cleaning of laboratory glassware and reusable laboratory material.
  • Good rinsing properties
  • Free from tensides
  • Removes limescale and other soils soluble in acids
ProCare Lab 30 P is intended for general use in medical, biological, microbiological and chemical laboratories, food processing, cosmetics and pharmaceutical industries as well as the mineral oil sector and other branches.
Note: Suitable for stainless steel, laboratory glassware, plastics such as PE, PP, PVDF, PTFE. Not suitable for use onMore Product Information