Detergents for glassware washers, ProCare Lab 10 AP
Lieferant: MIELE
10265640 10265620
MIEL10265640EA 0 CHF
MIEL10265640 MIEL10265620
Detergents for glassware washers, ProCare Lab 10 AP
Reinigungsmittel Waschmittel und Oberflächenreiniger
Liquid, alkaline detergent for machine based cleaning of laboratory glassware and laboratory utensils.
  • Removes stubborn or dried-on residues
  • Free from tensides, phosphates, oxidants
ProCare Lab 10 AP is intended for general use in medical, biological, microbiological and chemical laboratories, food-processing, cosmetics and pharmaceutical industries as well as the mineral oil sector and other branches.
Note: Suitable for stainless steel, laboratory glassware, ceramics, plastics such as PE, PP, PVDF, PTFE. Not suitable for use on non ferrous metals,More Product Information