Masterflex® I/P® High-Performance Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Lieferant: Avantor Fluid Handling
MFLX96511-88EA 783 CHF
MFLX96511-88 MFLX96510-70 MFLX96510-88 MFLX96510-89
Masterflex® I/P® High-Performance Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Schläuche Schläuche für Peristaltikpumpen
Reduce the risk of particulate entrapment with smooth inner surface.
  • Free Certificate of Compliance is available - request in cart
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique
More Product Information