Flowline Nonintrusive RF Capacitance Level Switch
Lieferant: Avantor Fluid Handling
43299-12
MFLX43299-12EA 354 CHF
MFLX43299-12
Flowline Nonintrusive RF Capacitance Level Switch
Füllstandssensoren
Measure level without touching fluid to avoid contamination.
  • Ideal for applications where purity is important
If you are concerned about your liquid's purity, then this nonintrusive level switch is the solution. Sensor mounts on the outside of your nonmetallic tank and never touches your liquid. Use with clean, noncoating, nonscaling, and conductive liquids. NEMA 4X polysulfone sensor enclosure is ideal for industrial environments. Includes 8-ft cable. Weld mounting bracket (sold separately) in place for firm hold.