Reducer swivel adapter for John Guest polypropylene fittings
Lieferant: Avantor Fluid Handling
MFLX34007-57EA 45.8 CHF
MFLX34007-57
Reducer swivel adapter for John Guest polypropylene fittings
Fittings und Verbinder
Quick, leakproof connection.
  • Polypropylene and EPDM materials meet NSF 51, and NSF 61 requirements for food-grade applications, and is FDA compliant
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semi-rigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper,More Product Information