CPC® Plastic Quick-Disconnect Fittings, Hose Barb Bodies
Lieferant: Avantor Fluid Handling
MFLX31311-12EA 28.4 CHF
MFLX31311-12 MFLX31311-13 MFLX31311-16 MFLX31311-17 MFLX31311-18 MFLX31311-21 MFLX31311-22 MFLX31311-23 MFLX31311-27 MFLX31311-28 MFLX06360-94 MFLX06360-95 MFLX31311-11 MFLX31311-26 MFLX31311-16BLK MFLX31311-21BLK MFLX06360-94BLK MFLX31311-27BLK MFLX31311-22BLK MFLX31311-18BLK MFLX31311-28BLK MFLX06360-95BLK MFLX31311-12BLK MFLX31311-26BLK MFLX31311-11BLK MFLX31311-13BLK MFLX31311-23BLK MFLX31311-17BLK
CPC® Plastic Quick-Disconnect Fittings, Hose Barb Bodies
Fittings und Verbinder
Fittings connect in a snap.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Acetal and ABS fittings are ideal for use with corrosive fluids
  • Polycarbonate and polysulfone materials meet USP Class VI requirements
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body.
AMore Product Information