Masterflex® Single-Use Flow Tube Sensors, Avantor®
Lieferant: Avantor Fluid Handling
MFLX24507-05EA 246 CHF
MFLX24507-05 MFLX24507-06 MFLX24507-10 MFLX24507-30 MFLX24507-31 MFLX24507-32 MFLX24507-01 MFLX24507-02 MFLX24507-03 MFLX24507-09 MFLX25507-44 MFLX24507-04 MFLX24507-07 MFLX24507-08 MFLX24507-33
Masterflex® Single-Use Flow Tube Sensors, Avantor®
Fluidsensoren
Build a complete single-use flow sensor system customizable to your application.
  • Modular interchangeable flow sensor systems with single-use design
  • Accurate 1% of reading measurement at low flow
  • Low operating cost disposable flow tubes for optimal contamination-free production
  • Ideal for single-use applications while also suitable for long-term measurement
  • Flow tubes and turbine sensors are produced and tested under cleanroom conditions
The Masterflex® single-use flow sensor system is comprised of four components. Select the flow sensor, flow tubes,More Product Information