Variable-Speed Drives for Micropump Low-Flow Pump Heads
Lieferant: Avantor Fluid Handling
07002-37 07002-38 07002-39
MFLX07002-37EA 3690 CHF
MFLX07002-37 MFLX07002-38 MFLX07002-39
Variable-Speed Drives for Micropump Low-Flow Pump Heads
Pumpen Pumpenantriebe Zahnradpumpenantriebe
Ideal for use with low flow pump heads.
  • Compatible with Micropump® low-flow C-mount pump heads without canisters (order separately)
  • Magnetic seal minimizes friction—all horsepower is transmitted directly to the motor gears
  • Hundreds of hours of dependable, leakproof service
  • Variable speed operation with optional power supply/speed controller—order at side.
Order drives at side; order power supply/speed controller separately at side. Power supply includes a 2-amp fuse and a 6-ft, UL-listed, three-wire cord with plug for 115 VAC operation and measure 7 3/More Product Information