Masterflex® B/T® PerfectPosition® Pumpenschläuche, PharMed® BPT, Avantor®
Lieferant: Avantor Fluid Handling
MFLX06507-87EA 231 CHF
MFLX06507-87 MFLX06507-91 MFLX06508-91 MFLX06508-90 MFLX06508-92
Masterflex® B/T® PerfectPosition® Pumpenschläuche, PharMed® BPT, Avantor®
Schläuche Schläuche für Peristaltikpumpen
Ensure optimum performance from your Masterflex® pump.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for
More Product Information