Super Erecta® Shelving Systems, Wire
Lieferant: Metro International
2460BR 1448NC 1842NS 1842NK3 1848NK3 1854NK3 1860NK3 1836NC 2448NC 2460NC 2448NS 1424BR 1436BR 1442BR 1448BR 1460BR 1854BR 2124BR 2472BR 2154BR 2136BR 2472NK3 1830NK3 1836NK3 1460NK3 1824NK3 2472NC 1842NC 1854NC 2436NS 2142NC 1824NC 1872NS 1848NS 2442NS 2442NK3 1824NS 1836BR 3648NC 2136NS 2136NC 2142NS 2148NC 1872NC 2148NS 2154NC 2154NS 2160NS 2172NS 1848NC 1854NS 1860NC 1860NS 1836NS 2454NC 2436NC 2442NC 2454NS 2460NS 2472NS 2160NC 2172NC 3060NC 3072NC 1460NC 1460NS 2130NC 1430NS 1472NS 1424NS 2130NS 1436NS 1424NC 1436NC 1818NC 1830NC 1430NC 2460NK3 1442NC 1472NC 2124NC 2124NS 2424NC 3048NS
METO2460BREA 299 CHF
METO2460BR METO1448NC 139-0297 METO1842NK3 METO1848NK3 METO1854NK3 METO1860NK3 139-0278 139-0292 139-0294 139-0314 METO1424BR METO1436BR METO1442BR METO1448BR METO1460BR METO1854BR METO2124BR METO2472BR METO2154BR METO2136BR METO2472NK3 METO1830NK3 METO1836NK3 METO1460NK3 METO1824NK3 139-0295 139-0279 139-0281 139-0312 139-0285 METO1824NC 139-0305 139-0298 139-0313 METO2442NK3 METO1824NS METO1836BR METO3648NC 139-0306 139-0284 139-0307 139-0286 139-0283 139-0308 139-0287 139-0309 139-0310 139-0311 139-0280 139-0299 139-0282 139-0304 139-0296 139-0293 139-0290 139-0291 139-0315 139-0316 139-0317 139-0288 139-0289 METO3060NC METO3072NC METO1460NC METO1460NS METO2130NC METO1430NS METO1472NS METO1424NS METO2130NS METO1436NS METO1424NC METO1436NC METO1818NC METO1830NC METO1430NC METO2460NK3 METO1442NC METO1472NC METO2124NC METO2124NS METO2424NC METO3048NS
Super Erecta® Shelving Systems, Wire
Möbel Regale
The open wire design of these heavy-gauge shelves minimizes dust accumulation and allows a free circulation of air, greater visibility of stored items and greater light penetration.
  • Fast, secure assembly: shelf is assembled in minutes without the use of any special tools
  • Versatile construction: by using various accessories, hundreds of shelving configurations become possible
  • Accessibility: shelves can be loaded/unloaded easily from all sides; this open construction allows use of maximum storage space of cube
  • Adjustability: shelves can be adjusted 25 mm
More Product Information