Avantor® ACE® ChromSword, HPLC/UHPLC Columns, Method Development Kits
Lieferant: Avantor
MDKCS-17-0502UEA 5110 CHF
MDKCS-17-0502U MDKCS-17-0503U MDKCS-17-1002U MDKCS-17-1003U MDKCS-2-0502U MDKCS-2-0503U MDKCS-2-0546U MDKCS-2-1002U MDKCS-2-1003U MDKCS-2-1046U MDKCS-2-1502U MDKCS-2-1503U MDKCS-2-1546U MDKCS-3-0502U MDKCS-3-0503U MDKCS-3-0546U MDKCS-3-1002U MDKCS-3-1003U MDKCS-3-1046U MDKCS-3-1502U MDKCS-3-1503U MDKCS-3-1546U MDKCS-3-2502U MDKCS-5-0502U MDKCS-5-0503U MDKCS-5-0546U MDKCS-5-1002U MDKCS-5-1003U MDKCS-5-1046U MDKCS-5-1502U MDKCS-5-1503U MDKCS-5-1546U MDKCS-5-2502U MDKCS-5-2503U MDKCS-5-2546U
Avantor® ACE® ChromSword, HPLC/UHPLC Columns, Method Development Kits
Chromatographiesäulen
Solve U/HPLC method development challenges systematically and efficiently using Avantor® ACE® columns and ChromSword 5.1 Lite method development software. The Avantor® ACE® ChromSword Method Development Kit includes six alternative selectivity Avantor® ACE® phases, a copy of ChromSword 5.1 Lite offline with a six month activation licence and a lot of technical information and support.
  • Ideal tool to investigate method development
  • Excellent efficiency, reproducibility and column lifetime
  • Columns with different mechanisms of interaction grouped in a kit to
More Product Information