5α-Cholestan

Lieferant: MATREYA

1115 1115
MAYA1115EA 0 CHF
MAYA1115 BTRN1115
5α-Cholestan
5α-Cholestan
Formel: C₂₇H₄₈
MW: 372.68 g/mol
Siedepunkt: 441
Melting Pt: 79
Dichte: 0.9133 g/cm³ (80 °C)
MDL Number: MFCD00066412
CAS-Nummer: 481-21-0
EINECS: 207-562-8
Merck Index: 12,02254

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR