Zellulose, Avicel

Lieferant: MACHEREY-NAGEL

Synonyms: Microcrystallinecellulose

815290.1 815290.5
MANA815290.1EA 79.1 CHF
MANA815290.1 MANA815290.5
Zellulose, Avicel
Zellulose
Formel: (C₁₂H₂₂O₁₁)n
Melting Pt: 260…270 °C
Dichte: 1.27…1.6 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00081512
CAS-Nummer: 9004-34-6
EINECS: 232-674-9
Merck Index: 13,01977

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR