Glass wool
Lieferant: MACHEREY-NAGEL
481100 281120 281130 481110
MANA481100EA 27.5 CHF
MANA481100 MANA281120 MANA281130 MANA481110
Glass wool
Glas- und Baumwolle
Filter aids transform difficult precipitates and colloidal particles into a form which can be easily filtered. When slimy and strongly lyophilic, swelling precipitates are involved, the fibres of the filter flocs prevent formation of a continuous, impermeable layer on the filter. The resulting filter cake remains porous and permeable, and clogging of the filter is prevented.
Order Now
TECHNISCHE DATEN
Environmentally Preferable