Florisil®, BAKER ANALYZED® 100 - 200 mesh, Laboratory Reagent Chromatography, J.T.Baker®

Lieferant: Avantor

Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil

M369-07EA 308 CHF
M369-07
Florisil®, BAKER ANALYZED® 100 - 200 mesh, Laboratory Reagent Chromatography, J.T.Baker®
Magnesiumsilikat

Selective adsorbent for gas chromatography. Contains activated magnesium silicate.

Formel: MgO₃Si
MW: 100.38 g/mol
Dichte: 2.51 Kg/L
MDL Number: MFCD00078249
CAS-Nummer: 1343-88-0
EINECS: 215-681-1

Order Now

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR