Calceinblau fluorescent dye, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyms: 4-Methylumbelliferonemethyleneiminodiacetic Acid, Umbelliferone Complexon

MQ200

M1255-10G M1255-1G
M1255-10GEA 505 CHF
M1255-10G M1255-1G
Calceinblau fluorescent dye, Sigma-Aldrich®
Calcein blue
Formel: C₁₅H₁₅NO₇
MW: 321.29 g/mol
Lager Temperatur: Room Temperature
MDL Number: MFCD00006867
CAS-Nummer: 54375-47-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c13ed38ed164fc731492db6a7ab81e94a6f9f457&vid=e4afe27e42b6be7a8756f76824de17af3e23ecb8&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance (Colour) White to Light Yellow
Appearance (Form) Powder
Solubility (Colour) Light Yellow to Dark Yellow and Light Orange to Dark Orange
Solubility (Turbidity)
50 mg/mL, 1 M NaOH
Clear to Slightly Hazy
Carbon 54.5 - 56.5 %
Nitrogen 4.0 - 4.6 %
Suitability
for use as an indicator for EDTA titrations
Suitable
1H NMR Spectrum Conforms to Structure

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR