Magnesiumsulfat

Lieferant: G Bioscienes

Synonyms: MgSO4, Magnesium sulphate anhydrous

M006
M006EA 226 CHF
M006
Magnesiumsulfat
Magnesium sulphate
Formel: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1127
Dichte: 2.65 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011110
CAS-Nummer: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR