DNA markers
Lieferant: LONZA
50461 50471
LONZ50461EA 516 CHF
LONZ50461 LONZ50471
DNA markers
Elektrophorese-Reagenzien Molekulargewichtsmarker
DNA marker is for rapid size estimation of PCR products and restriction fragments. DNA can be visualised by ethidium bromide staining or kinased with radiolabelled ³²P for detection by autoradiography.